Learn More
PEX14 (Human Specific) Rabbit anti-Human, Unconjugated, Clone: 6K5, Proteintech
Código de producto. 17422453 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
17422453 20 μL 20 microlitros
17412453 100 μL 100 microlitros
2 options
Código de producto. 17422453
Marca: Proteintech 801961RR20UL
Rabbit Recombinant Antibody
The PEX14 (human specific) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PEX14 (human specific). This antibody recognizes human antigen. The PEX14 (human specific) antibody has been validated for the following applications: IF, WB, ELISA analysis.
Especificaciones
Antígeno PEX14 (Human Specific)
Aplicaciones Western Blot, Immunofluorescence
Clasificación Monoclonal
Clon 6K5
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 5195
Especies diana Human
Isotype IgG