Learn More
DiversifiedBiotech PetriStickers™
GREENER_CHOICE
Código de producto. p-200132132
Change view
Click to view available options
Incluye:
100 Square Grid
12 Sector Pie
16 Sector Pie
32 Square Grid
50 Square Grid
6 Sector Pie
70 Square Grid
8 Sector Pie
Tamaño de la unidad:
paquete de 36
Código de producto. Incluye unitSize
30282323 50 Square Grid paquete de 36
30282301 32 Square Grid paquete de 36
30282330 70 Square Grid paquete de 36
30282337 100 Square Grid paquete de 36
30282281 6 Sector Pie paquete de 36
30282289 8 Sector Pie paquete de 36
30282343 12 Sector Pie paquete de 36
30282342 16 Sector Pie paquete de 36
8 options
Código de producto. 30282323
Marca: DiversifiedBiotech PSTK1000
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Allow for petri dishes to be labeled quickly, neatly and reliably
  • PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably
  • Made with water-resistant ink, substrate and adhesives and will remain firmly attached at temperatures from 42° to 4°C
  • Simply remove the label's protective backing and apply it to the back of a petridish
  • After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed
Especificaciones
Tipo Petri Dish Label
Incluye 50 Square Grid
Diámetro (métrico) 7.62 cm