Learn More
DiversifiedBiotech PetriStickers™
Descripción
- PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably
- Made with water-resistant ink, substrate and adhesives and will remain firmly attached at temperatures from 42° to 4°C
- Simply remove the label's protective backing and apply it to the back of a petridish
- After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed
Especificaciones
Especificaciones
| Tipo | Petri Dish Label |
| Incluye | 100 Square Grid |
| Diámetro (métrico) | 7.62 cm |