Learn More
Peroxiredoxin 2 Mouse anti-Human, CoraLite 488, Clone: 3H6C4, Proteintech
Tienda Proteintech ProductosDescripción
The peroxiredoxin 2 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human peroxiredoxin 2. This antibody recognizes human, mouse antigen. The peroxiredoxin 2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Especificaciones
Especificaciones
| Antígeno | Peroxiredoxin 2 |
| Aplicaciones | Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | 3H6C4 |
| Concentración | 2000 μg/mL |
| Conjugado | CoraLite 488 |
| Especie del huésped | Mouse |
| Cantidad | 100 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 7001 |
| Mostrar más |