Learn More
Liebherr™ Performance High-Volume Upright Lab Fridge with Stainless Steel Interior
GREENER_CHOICE
Código de producto. p-200089300
Change view
Click to view available options
Capacidad (métrico):
480 L
702 L
1091 L
Tamaño de la unidad:
cada uno
Código de producto. Capacidad (métrico) unitSize
30178708 1091 L cada uno
30178701 480 L cada uno
30178692 702 L cada uno
3 options
Código de producto. 30178708
Marca: Liebherr™ SRPVG1402
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Upright refigerator designed for lab applications that provides secure, accurate and energy-efficient fridge storage of temperature-sensitive products.
  • High-volume upright lab fridge, reach-in
  • Temperature range -2 to +16°C, ± 3 K temperature stability tested to EN60068-3
  • White exterior, stainless steel interior, fan-assisted refrigeration.
  • Refrigerant R 290, connection rating 4.0 A, 220-240V/50/1-ph
  • Energy-efficiency optimised for lower running costs and help to meet user sustainability targets
  • Digital controller, high-visibility monochrome display with touch buttons, 0.1°C resolution provides useful extended multi-functionality, e.g. a maintenance timer, calibration function et al.
  • Connectivity: LAN/WLAN interface not integrated but retrofittable as accessory
  • 12 mm access port for external temperature probe
  • Content security: hi/low temperature alarms and power failure alarm when power supply returns.
  • Ceiling-mounted LED internal lighting for improved visibility of the contents of this large appliance
  • Mechanical door locks, reversible hinges, self-closing doors with 90° dwell position
  • 60 kg shelf load
  • Equipped with 100 mm castors, with brakes on the two front, for ease of moving the unit
Especificaciones
Tipo Refrigerador
Capacidad (imperial) 288.21 gal.
Capacidad (métrico) 1091 L
Intervalo de temperatura -2°C to +16°C
Resolución de temperatura 0.1 K
Estabilidad de la temperatura ±3°K
Refrigerante R 290
Modelo de puerta Solid Reversible Self-closing with Lock
No. of Doors 2
Lado abisagrado Left or Right
Estantes 8
Material de la cabina Acero pintado resistente a los arañazos
Acabado exterior White Powder-coated
Certificaciones/conformidad ISO 9001, ISO 14001, CE
Modelo Vertical
Pantalla High-Contrast Monochrome Display with Touch Buttons
Ruedas giratorias 100 mm dia. castors
Color White
Tipo de condensador Low-Maintenance Wire-on-Tube with Washable Filter
Tipo de control Digital Controller
Puertos 1 x 12 mm dia. access port
Garantía 2 Years Parts and Labor
Descongelación Automatic Hot-Gas Defrost
Iluminación LED Ceiling Mounted
Opciones de vigilancia SmartCoolingHub
Seguridad de acceso Mechanical Door Lock with Key
Conectividad WiFi/Lan Module Optional
compresor CFC-free
Amperaje 4 A
tipo de tapón CEE 7/7
Tensión 220/240 V
Frecuencia 50 Hz
Fase 1, Single Phase
Consumo energético 464 kWh/a
Características ecológicas Natural Refrigerant, Super Energy Efficient, Ecovadis Gold