missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Placas matriz de agujeros para desecador

Código de producto. 10112541 Tienda Bürkle Productos
Change view
Click to view available options
Diámetro interior (métrico):
200 mm
250 mm
Unit Size:
cada uno
Product Code. Diámetro interior (métrico) unitSize
10112541 200 mm cada uno
10784004 250 mm cada uno
2 options
This item is not returnable. View return policy

Product Code. 10112541

Brand: Buerkle™ 97910201

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pieza insertada agujereada de polipropileno de Buerkle™.

  • Desiccator plate
  • Made out of Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Diámetro exterior (imperial) 190 mm
Tipo de producto Placa matriz de agujeros
Material Polipropileno
Para utilizar con (equipo) Desecador
Diámetro interior (métrico) 200 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.