missing translation for 'onlineSavingsMsg'
Learn More

Pepsinogen C/PGC/Progastricsin Antibody (90/11), CoraFluor™ 1, Novus Biologicals™

Product Code. 30150440
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30150440 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30150440 Supplier Novus Biologicals (Bio-Techne) Supplier No. NB10062819CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pepsinogen C/PGC/Progastricsin Monoclonal antibody specifically detects Pepsinogen C/PGC/Progastricsin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Pepsinogen C/PGC/Progastricsin
Aplicaciones ELISA
Clasificación Monoclonal
Clon 90/11
Conjugado CoraFluor 1
Formulación PBS
Alias de gen EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C)
Especie del huésped Mouse
Inmunógeno Human pepsinogen II
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 5225
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.