Learn More
Penicillin G sodium salt, Thermo Scientific™
Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria.
Marca:  Thermo Scientific Alfa Aesar J63032.22
Código de producto. 15405309
  • 150.00€ / 100 g
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
• Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis
Applications
• Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
• It can also be used to prevent infection in research animals
Videos
SDS Documentos