Learn More
Invitrogen™ PEN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5144453
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PEN1 Polyclonal Antibody for Western Blot, ELISA
Converts oxidosqualene to arabidiol. Minor production of arabidiol 20,21-epoxide.
Especificaciones
| PEN1 | |
| Polyclonal | |
| Unconjugated | |
| PEN1 | |
| 04C11 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 827200 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.5 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q9FR95 | |
| PEN1 | |
| Recombinant Arabidopsis thaliana PEN1 protein. | |
| 100 μL | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |