Learn More
Gilac™ Pedal Container with Lid, 50 L
Descripción
- Compact pedal container for space saving.
- Hands-free operation to prevent contamination and facilitate barrier gestures, pedal prevents contamination.
- 2 pedal mounting positions available: automatic or manual lid closing.
- The lid (in manual lid version) can be closed by pressing the left part of the pedal.
- Longevity and durability.
- Perfect stability.
- Easy to clean.
- Colored lids for selective waste sorting.
- Self-assembled kit: lid, body, screws and pedal delivered in the same box.
Especificaciones
Especificaciones
| Capacidad (métrico) | 50 L |
| Capacidad (imperial) | 13.2 gal. |
| Color | Amarillo |
| Esterilizables en autoclave | Not Autoclavable |
| Material | Polypropylene |
| Tipo de producto | Pedal Container with Lid |