missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDZD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA521886
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: A549. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Proteins containing PDZ domains have been shown frequently to bind the C-termini of transmembrane receptors or ion channels. They have also been shown to bind to other PDZ domain proteins and could possibly be involved in intracellular signaling. The protein encoded by this gene contains six PDZ domains and shares sequence similarity with pro-interleukin-16 (pro-IL-16). Like pro-IL-16, the encoded protein localizes to the endoplasmic reticulum and is thought to be cleaved by a caspase to produce a secreted peptide containing two PDZ domains. In addition, this gene is upregulated in primary prostate tumors and may be involved in the early stages of prostate tumorigenesis.
Especificaciones
| PDZD2 | |
| Polyclonal | |
| Unconjugated | |
| PDZD2 | |
| 4930537L06Rik; A930022H17Rik; activated in prostate cancer protein; AIPC; Gm82; KIAA0300; mKIAA0300; PAPIN; PDZ domain containing 2; PDZ domain containing 3; PDZ domain-containing protein 2; PDZ domain-containing protein 3; PDZD2; PDZK3; Pin1; Plakophilin-related armadillo repeat protein-interacting PDZ protein; plakophilin-related armadillo repeat protein-interactive PDZ protein; Processed PDZ domain-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23037 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O15018 | |
| PDZD2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PDZK3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido