missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDLIM1 Recombinant Rabbit Monoclonal Antibody (24GB1310)

Código de producto. 30275847
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
Código de producto. Cantidad unitSize
30275847 20 μL 20 microlitros
30275607 100 μL 100 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30275847

Marca: Invitrogen™ MA557551

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

PDLIM1 Recombinant Monoclonal Antibody for Western Blot, Flow

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. A distinctive feature of TBP is a long string of glutamines in the N-terminal. This region of the protein modulates the DNA binding activity of the C terminus, and modulation of DNA binding affects the rate of transcription complex formation and initiation of transcription. Mutations that expand the number of CAG repeats encoding this polyglutamine tract, and thus increase the length of the polyglutamine string, are associated with spinocerebellar ataxia 17, a neurodegenerative disorder classified as a polyglutamine disease.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PDLIM1
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB1310
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica Pdlim1
N.º de referencia del gen O00151, O70400, P52944
Alias de gen carboxyl terminal LIM domain protein 1; Clim1; Clp36; CLP-36; C-terminal LIM domain protein 1; elfin; enigma; epididymis secretory protein Li 112; hCLIM1; HEL-S-112; LIM domain protein CLP-36; LIM protein; mClim1; Pdlim1; PDZ and LIM domain 1; PDZ and LIM domain 1 (elfin); PDZ and LIM domain protein 1
Símbolos de los genes Pdlim1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human PDLIM1 (aa 201-265).
Método de purificación Affinity chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 54132, 54133, 9124
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.