Learn More
PDI Rabbit, anti-Human, Unconjugated, Clone: 4K17, Proteintech
Código de producto. 17241546 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17241546 20 μL 20 microlitros
17231546 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17241546 Proveedor Proteintech N.º de proveedor 801701RR20UL
Rabbit Recombinant Monoclonal Antibody
The PDI antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human PDI. This antibody recognizes human, mouse, rat antigen. The PDI antibody has been validated for the following applications: IF, IHC, WB,ELISA analysis.
Especificaciones
Antígeno PDI
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Monoclonal
Clon 4K17
Concentración 500 μg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 5034
Especies diana Human, Mouse, Rat