missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDI Monoclonal Antibody (2F6G12H2), Invitrogen™

Código de producto. p-7240411
Change view
Click to view available options
Cantidad:
50 μL
100 μL
Unit Size:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
17789743 50 μL 50 microlitros
17789553 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17789743 Supplier Invitrogen™ Supplier No. MA543390

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human P4HB/ERBA2L

Protein disulfide isomerase (PDI) is a multifunctional protein that catalyzes the formation, breakage and rearrangement of disulfide bonds. At the cell surface, the protein seems to act as a reductase that cleaves disulfide bonds of proteins attached to the cell. It may therefore cause structural modifications of exofacial proteins. Inside the cell, PDI seems to form/rearrange disulfide bonds of nascent proteins. At high concentrations, it functions as a chaperone that inhibits aggregation of misfolded proteins. At low concentrations, it facilitates aggregation (anti-chaperone activity). PDI may be involved with other chaperones in the structural modification of the TG precursor in hormone biogenesis. It also acts a structural subunit of various enzymes such as prolyl 4-hydroxylase and microsomal triacylglycerol transfer protein MTTP. Receptor for LGALS9; the interaction retains P4HB at the cell surface of Th2 T helper cells, increasing disulfide reductase activity at the plasma membrane, altering the plasma membrane redox state and enhancing cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno PDI
Aplicaciones ELISA, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 2F6G12H2
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica P4hb
N.º de referencia del gen P07237
Alias de gen 1810041F13Rik; AI661267; CaO19.12595; cellular thyroid hormone binding protein; cellular thyroid hormone-binding protein; CLCRP1; collagen prolyl 4-hydroxylase beta; DSI; Endoplasmic reticulum resident protein 59; Eps1p; ER protein 59; ERBA2L; ERp59; GIT; glutathione-insulin transhydrogenase; I79_002399; LOW QUALITY PROTEIN: protein disulfide-isomerase A2; MFP1; P4HB; p4hb.L; P4Hbeta; p55; p58; Pancreas-specific protein disulfide isomerase; pancreatic protein disulfide isomerase; PDA2; PDI; PDI (E.C.5.3.4.1); PDI1; PDI1P; Pdia1; pdia2; pdia2.L; PDIP; Pdipl; PDIR; PHDB; PO4DB; PO4HB; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), beta polypeptide; PROHB; prolyl 4-hydroxylase beta polypeptide; Prolyl 4-hydroxylase subunit beta; prolyl 4-hydroxylase subunit beta L homeolog; prolyl 4-hydroxylase subunit beta L homeolog; prolyl 4-hydroxylase, beta polypeptide L homeolog; prolyl 4-hydroxylase, beta polypeptide; prolyl 4-hydroxylase, beta subunit; protein disulfide isomerase; protein disulfide isomerase (pancreas) like; Protein disulfide isomerase (Prolyl 4-hydroxylase, beta polypeptide); protein disulfide isomerase A2; protein disulfide isomerase associated 2; protein disulfide isomerase family A member 2; protein disulfide isomerase family A member 2 L homeolog; protein disulfide isomerase family A, member 1; protein disulfide isomerase family A, member 2; protein disulfide isomerase PDI1; protein disulfide isomerase, pancreatic; protein disulfide isomerase/oxidoreductase; protein disulfide isomerase-associated 1; protein disulfide isomerase-associated 2; protein disulfide-isomerase; Protein disulfide-isomerase A2; protocollagen hydroxylase; Rho GDP dissociation inhibitor gamma; testicular secretory protein Li 32; Thbp; Thioredoxin-related glycoprotein 1; thyroid hormone-binding protein p55; TRG1; XELAEV_18043753mg; XELAEV_18045582mg; XPDIp; YCL043C; YCL313; YCL43C
Símbolos de los genes P4hb
Especie del huésped Mouse
Inmunógeno Recombinant Human P4HB protein.
Método de purificación Protein A
Cantidad 50 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5034
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.