Learn More
PDGFRB(phospho Y771) & PDGFRB Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Marca: Abnova DP0106.1Set
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein phosphorylation antibody pair set comes with two antibodies, one against the PDGFRB protein, and the other against the specific Y771 phosphorylated site of PDGFRB for use in in situ Proximity Ligation Assay.Especificaciones
| PDGFRB | |
| Unconjugated | |
| CD140B/JTK12/PDGF-R-beta/PDGFR/PDGFR1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-PDGFRB Y771 rabbit polyclonal antibody (20 ul) 2. PDGFRB mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| platelet-derived growth factor receptor, beta polypeptide | |
| PDGFRB | |
| RUO | |
| 5159 | |
| Human | |
| Ab Pair |