Learn More
PDE4B Monoclonal Antibody (OTI2B10), TrueMAB™, OriGene
Código de producto. 17539301
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
17539301 100 μL 100 microlitros
17559301 30 μL 30 microlitros
2 options
Código de producto. 17539301
Marca: Origene Technologies TA503446
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
Mouse Monoclonal Antibody
This gene is a member of the type IV, cyclic AMP (cAMP)-specific, cyclic nucleotide phosphodiesterase (PDE) family. Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. The cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. This gene encodes a protein that specifically hydrolyzes cAMP. Altered activity of this protein has been associated with schizophrenia and bipolar affective disorder. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Especificaciones
Antígeno PDE4B
Aplicaciones Flow Cytometry, Western Blot
Clasificación Monoclonal
Clon OTI2B10
Concentración 0.53 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica PDE4B
N.º de referencia del gen Q07343
Alias de gen DPDE4, PDEIVB
Símbolos de los genes PDE4B
Especie del huésped Mouse
Inmunógeno Full length human recombit protein of human PDE4B produced in HEK293T cell.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5142
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1