missing translation for 'onlineSavingsMsg'
Learn More

PDCD2L Monoclonal Antibody (OTI4H4), TrueMAB™, OriGene

Código de producto. 17596894
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17596894 30 μL 30 microlitros
17576894 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. View return policy
Product Code. 17596894 Supplier Origene Technologies Supplier No. TA812457S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDCD2L Monoclonal antibody specifically detects PDCD2L in Human samples. It is validated for Western Blot

Specifications

Antígeno PDCD2L
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI4H4
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica PDCD2L
N.º de referencia del gen Q9BRP1
Alias de gen 6030457N17Rik; MGC13096; PDCD2L; programmed cell death 2 like; programmed cell death 2-like; programmed cell death protein 2-like; RGD1308249
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human PDCD2L produced in HEK293T cell.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 84306
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.