missing translation for 'onlineSavingsMsg'
Learn More

PD-L2 (PDCD1LG2) Mouse anti-Human, Clone: UMAB223, liquid, UltraMAB™

Código de producto. 17569954
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
17569954 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 17569954

Brand: Origene Technologies UM800115

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD273 (PDCD1 Ligand 2, Programmed cell death 2 ligand 2, PD-L2, B7-DC) is involved in cell-mediated immune responses initiated by T lymphocytes that are stimulated by cognate peptides bound to MHC molecules on antigen-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as CD273 that mediate inhibitory signals from the APC (antigen presenting cell). CD273 can bind two different but related ligands, PDL-1 and PDL-2, both of which are thought act as a negative regulator of T cell activation. However, it has been suggested that PDL-2 can act to stimulate an immunogenic response through and alternative receptor from PD-1. At least three isoforms of PDL-2 are known to exist; CD273 is specific to the largest isoform. Diseases associated with CD273 include cysticercosis, and myasthenic syndrome.

Specifications

Antígeno PD-L2 (PDCD1LG2)
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon UMAB223
Concentración 0.5-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica PDCD1LG2
N.º de referencia del gen Q9BQ51
Alias de gen B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2
Símbolos de los genes PDCD1LG2
Especie del huésped Mouse
Inmunógeno Full length human recombit protein of human PDCD1LG2 produced in HEK293T cell.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 80380
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.