Learn More
PD-L1/CD274 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16897964 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16897964 20 μL 20 microlitros
16887964 150 μL 150 microlitros
2 options
Código de producto. 16897964
Marca: Proteintech 179521AP20UL
Rabbit Polyclonal Antibody
PD-L1 (Programmed death receptor ligand 1, B7-H1, CD274) is a 40 kDa transmembrane protein and B7 family member. PD-L1 has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production, and PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion. Together with B7.1, PD-L1 can inactivate T cells and this relationship is a focal point of cancer therapeutic treatment. At least two isoforms of PDL-1 are known to exist.
Especificaciones
Antígeno PD-L1/CD274
Aplicaciones Immunofluorescence, Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.33 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica CD274
N.º de referencia del gen Q9EP73, Q9NZQ7
Alias de gen PD-L1/CD274
Símbolos de los genes CD274
Especie del huésped Rabbit
Inmunógeno PD-L1/CD274 Fusion Protein Ag12432
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 29126, 499342, 60533
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG