missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCSK9 Monoclonal Antibody (15), Invitrogen™

Mouse Monoclonal Antibody
Marca: Invitrogen MA542994
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human PCSK9
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
Especificaciones
| PCSK9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8NBP7 | |
| PCSK9 | |
| Recombinant Human PCSK9 Protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 15 | |
| Unconjugated | |
| PCSK9 | |
| AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 | |
| Mouse | |
| Protein A | |
| RUO | |
| 255738 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido