missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCSK5 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15734541
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15734541 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15734541

Proveedor: Invitrogen™ PA556631

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: YLEHVVVRIT ITHPRRGDLA IYLTSPSGTR SQLLANRLFD HSMEGFKNWE FMTIHCWGER AAGDWVLEVY DTPSQLRNFK TPGKLKEWSL VLYGTSVQPY SPTNEFPKVE RFRYSRVEDP TDD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

PCSK5 belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. PCSK5 mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known. The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PCSK5
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Polyclonal
Concentración 0.1 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica Pcsk5
N.º de referencia del gen Q92824
Alias de gen b2b1549Clo; b2b585Clo; PC5; PC5/6A; PC5A; PC6; PC6A; PCSK5; prohormone convertase 5; proprotein convertase 5; Proprotein convertase 6; proprotein convertase PC5; proprotein convertase subtilisin/kexin type 5; protease PC6; rPC5; SPC6; subtilase; Subtilisin/kexin-like protease PC5; subtilisin-like proprotein convertase 6
Símbolos de los genes Pcsk5
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human PCSK5. Recombinant protein control fragment (Product #RP-89239).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5125
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.