Learn More
PCNA (Proliferating Cell Nuclear Antigen), Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Código de producto. 12613427 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
500 μL
1 mL
7 mL
Formulario:
Concentrated
Prediluted
Tamaño de la unidad:
0.10 ml
1 ml
500 microlitros
7 ml
Código de producto. Cantidad Formulario unitSize
12613427 500 μL Concentrated 500 microlitros
12603427 100 μL Concentrated 0.10 ml
12693417 1 mL Concentrated 1 ml
12643427 7 mL Prediluted 7 ml
4 options
Código de producto. 12613427
Marca: Epredia RB9055P1
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Provide accurate, reproducible results with the Epredia™ PCNA (Proliferating Cell Nuclear Antigen), Rabbit Polyclonal Antibody.
Expression of proliferating cell nuclear antigen (PCNA) or cyclin or polymerase delta auxiliary protein is elevated in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. PCNA/cyclin may act as an auxiliary protein of DNA polymerase-delta to play a fundamental role in the initiation of cell proliferation.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant human PCNA protein
Molecular Weight: 36kDa
Positive Control: Raji cells, Tonsil, Lymph node, and small intestine
Cellular Localization: Nuclear
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PCNA (Proliferating Cell Nuclear Antigen)
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 200µg/mL
Conjugado Unconjugated
Especie del huésped Rabbit
Inmunógeno Recombinant human PCNA protein
Cantidad 500 μL
Estado normativo IVD
Disciplina de investigación Cell Cycle
Primario o secundario Primary
Especies diana Human
Formulario Concentrated
Isotype IgG