Learn More
PCNA Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Novus Biologicals NB100-456C
Código de producto. 18025234
  • 639.00€ / 100 microlitros
Descripción
PCNA Polyclonal specifically detects PCNA in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
PCNA
Polyclonal
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Rabbit
Affinity Purified
RUO
Primary
Human, Mouse
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
PCNA
The immunogen this antibody was made to, maps to a region between residues 75-125 of human proliferating cell nuclear antigen using the SwissProt entry P12004 (GeneID 5111).
100 μL
Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers
5111
Store at 4C in the dark.
Certificados
Promociones
Promociones
For Research Use Only