Learn More
DiscoverX pCMV- ProLinkTM 2 Vector
Código de producto. 17312679
Encompass
Change view
Click to view available options
Cantidad:
1 vector
Tamaño de la unidad:
1 set
17312679 1 vector 1 set
1 options
Código de producto. 17312679
Marca: DiscoverX 930171
The pCMV- ProLinkTM 2 Vector is a mammalian cloning vector to produce cells expressing a PK2-tagged protein of interest. Provided as a vial of 10μg vector in liquid buffer.
The pCMV- ProLinkTM 2 Vector is a mammalian cloning vector for use in expressing a protein of interest fused to the ProLink 2 (PK 2) tag at the C-terminus. The PK tag is a version of ED used for EFC assays (in combination with EA). The ProLink 2 tag has a higher affinity for EA, making it ideal for use with weaker protein:protein interactions. Provided as a vial of 10μg vector in 20μL liquid buffer.
Especificaciones
Tipo de producto Cloning Vector
Cantidad 1 vector
Para utilizar con (aplicación) Cloning Vectors