missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCGF6 Polyclonal Antibody

Descripción
PCGF6 Polyclonal Antibody for Western Blot
Polycomb group (PcG) proteins form multiprotein complexes that regulate expression patterns of developmental and cell proliferation genes. Several members of the PcG contain ring finger domains and are identified as a subclass of RING finger proteins. The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain the RING-type zinc finger conserved domain are generally involved in the ubiquitination pathway of protein degradation. PCGF6 (polycomb group ring finger 6), also known as MBLR or RNF134, is a 350 amino acid nuclear protein that is ubiquitously expressed and contains one RING-type zinc finger. PCGF6 acts as a transcriptional repressor and regulates the level of histone H3K4Me3 by activating SmcY histone demethylase. PCGF6 exists as two alternatively spliced isoforms.
Especificaciones
Especificaciones
| Antígeno | PCGF6 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | PCGF6 |
| N.º de referencia del gen | Q5XI70, Q9BYE7 |
| Alias de gen | 4933407A11Rik; AI604840; Mblr; Mel18 and Bmi1-like RING finger; Mel18- and Bmi1-like RING finger protein; Mel18 and Bmi1-like RING finger protein; Pcgf6; polycomb group ring finger 6; polycomb group RING finger protein 6; RING finger protein 134; RNF134 |
| Símbolos de los genes | PCGF6 |
| Especie del huésped | Rabbit |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?