missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHGC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA531045
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: NT2D1, MCF-7. Predicted reactivity: Mouse (92%), Rat (93%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Especificaciones
| PCDHGC5 | |
| Polyclonal | |
| Unconjugated | |
| PCDHGC5 | |
| PCDH-gamma-C5; PCDHGC5; protocadherin gamma c5; protocadherin gamma subfamily C, 5; protocadherin gamma-C5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56097 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.75 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y5F6 | |
| PCDHGC5 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human PCDHGC5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido