missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHB6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA556192
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: QVPENNPLGS LVITVSARDL DAGSFGKVSY ALFQVDDVNQ PFEINAITGE IRLRKALDFE EIQSYDVDVE ATDGGGLSGK CSLVVRVLDV NDNAPELTMS FFISLIPENL PEITVAVFSV SDADS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.
This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections.
Especificaciones
| PCDHB6 | |
| Polyclonal | |
| Unconjugated | |
| PCDHB6 | |
| PCDHB6; PCDH-BETA6; PCDH-beta-6; POLR1E; protocadherin beta 6; protocadherin beta-6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56130 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5E3 | |
| PCDHB6 | |
| Recombinant protein corresponding to Human PCDHB6. Recombinant protein control fragment (Product #RP-90934). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido