missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-93346-0.1ml
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PBX2 Polyclonal antibody specifically detects PBX2 in Mouse, Rat samples. It is validated for Western Blot
Especificaciones
| PBX2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| G17Homeobox protein PBX2, HOX12homeobox 12, PBX2MHC, pre-B-cell leukemia homeobox 2, pre-B-cell leukemia transcription factor 2, Protein G17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 321-430 of human PBX2 (NP_002577.2). AVSVTQGGHSRTSSPTPPSSAGSGGSFNLSGSGDMFLGMPGLNGDSYSASQVESLRHSMGPGGYGDNLGGGQMYSPREMRANGSWQEAVTPSSVTSPTEGPGSVHSDTSN | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5089 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Spot an opportunity for improvement?Share a Content Correction