missing translation for 'onlineSavingsMsg'
Learn More

Melford PBS/Tween 20 10X Solution

Código de producto. 30018676
Change view
Click to view available options
Cantidad:
500 mL
Tamaño de la unidad:
500 ml
Código de producto. Cantidad unitSize
30018676 500 mL 500 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30018676

Marca: Melford P21200500.0

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

PBS/Tween 20 10X Solution is a widely used buffer solution in the life science marketplace. It is a combination of phosphate-buffered saline (PBS) and Tween 20, a nonionic detergent.

PBS/Tween 20 10X Solution is primarily utilized for various applications such as immunohistochemistry, Western blotting, and antibody-based assays. It serves as a buffer for sample dilution, antibody washing, and blocking steps during these experimental procedures.

The solution is prepared at a 10X concentration, meaning it is ten times more concentrated than the final desired concentration. This concentrated form allows for efficient storage and transportation, as well as flexibility in adjusting the final working concentration.

PBS is a buffer commonly used in biological and biochemical research. It provides a stable pH environment and maintains osmolarity, ensuring optimal conditions for biological reactions.

PBS is also compatible with biological samples, making it suitable for a wide range of applications. Tween 20, a nonionic detergent, is added to the PBS solution to reduce nonspecific binding and improve the efficiency of antibody-antigen interactions. It also helps in washing steps by reducing background noise and enhancing signal detection.

Especificaciones

Nombre del producto químico o material PBS/Tween 20 10 x Solution
CAS 7558-79-4, 7778-77-0, 7647-14-5, 7447-40-7, 9005-64-5, 7732-18-5
Cantidad 500 mL
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.