missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pazopanib Hydrochloride, MedChemExpress
Descripción
Pazopanib Hydrochloride (GW786034 Hydrochloride) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRβ, c-Kit, FGFR1, and c-Fms with an IC50 of 10, 30, 47, 84, 74, 140 and 146 nM, respectively.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Pazopanib Hydrochloride |
| CAS | 635702-64-6 |
| Color | Blanco |
| Notas de grado de pureza | Research |
| Fórmula molecular | C21H24ClN7O2S |
| Cantidad | 50 mg |
| Sinónimo | GW786034 (Hydrochloride) |
| Información de solubilidad | DMSO : 10 mg/mL (21.10 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | CC1=CC=C(C=C1S(N)(=O)=O)NC2=NC=CC(N(C3=CC4=NN(C(C)=C4C=C3)C)C)=N2.Cl |
| Peso molecular (g/mol) | 473.98 |
| Mostrar más |
Research purposes only
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?