Learn More
Invitrogen™ PAX5 Polyclonal Antibody

Descripción
Western blot analysis of PA1-109 detects a prominent ~45 kDa protein in PAX5 expressing human Ramos, Daudi, Gaji and mouse A20 B-cell lymphoma cells. No PAX5 was detected in negative control Jurkat cells. The 45 kDa band is enriched in immunoprecipitations from Raji lysate, however additional unknown high molecular weight bands are also enriched and detected by PA1-109. PAX5 was also specifically detected by immunofluorescence in the nuclei of Ramos cells compared to negative control Jurkat cells.
Especificaciones
Especificaciones
| Antígeno | PAX5 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide |
| génica | PAX5 |
| N.º de referencia del gen | Q02548, Q02650 |
| Alias de gen | ALL3; B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; EBB-1; KLP; paired box 5; paired box gene 5; paired box gene 5 (B-cell lineage specific activator); paired box homeotic gene 5; paired box protein Pax-5; paired domain gene 5; Pax5; Pax-5; Pax5 variant 1; Pax5 variant 2; RGD1565236; transcription factor PAX 5 |
| Símbolos de los genes | PAX5 |
| Mostrar más |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.