Learn More
Intelsius™ PathoShield™
Descripción
- Complete Category B shipping solution includes flexible 95kPa tested compliant secondary, absorbent, labels, and relevant UN markings
- Range of sizes suitable for shipping through postal and courier networks
- Suitable for a wide range of primaries including blood vials, swabs and sample containers
- Supplied flat packed for efficient storage and shipping prior to use
Especificaciones
Especificaciones
| Longitud (imperial) exterior | 7.5 in. |
| Anchura exterior (imperial) | 5.7 in. |
| Incluye | Pouch |
| Tamaño | 8 |
| Altura exterior (imperial) | 0.9 in. |
| Tipo | Sample Transport Packaging |
| Dimensiones (L x An) | Pouch: 150 x 190 mm (5.9 x 7.4 in.) |
| Dimensions (LxWxH) | 191 x 145 x 22 mm |