Learn More
DiscoverX PathHunter™ U2OS TrkA Functional Assay
Descripción
The PathHunter™ U2OS TrkA Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated TrkA dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged TrkA and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Especificaciones
Especificaciones
| Entrada del ensayo | Receptor Activation |
| Línea de células | U2OS |
| Para utilizar con (aplicación) | Cell Lines |
| Agonista primario | Recombinant Human b-NGF |
| Cantidad | 2 viales |
| Entrada diana | TrkA |
| Número de acceso | NM_002529 |