Learn More
DiscoverX PathHunter™ U2OS OSMRb/IL31R Bioassay Kit
Descripción
The PathHunter™ OSMRb/IL31R Dimerization Bioassay kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This bioassay assesses ligand (e.g. IL-31) induced dimerization of OSMRb/IL31R receptors. This product uses EFC technology. The cell line included overexpresses OSMRb target protein fused to PK alongside IL31RA target protein fused to EA. Upon ligand-induced dimerization of OSMRb/IL31R , detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | Dimerization |
| Para utilizar con (aplicación) | Assay-Ready Bioassay Kits |
| Cantidad | 1000 dp (10 x 96-pocillos) |
| Entrada diana | OSMRb/IL31R |
| Línea de células | U2OS |
| Objetivo | OSMRb/IL31R |