Learn More
DiscoverX PathHunter™ eXpress PTGIR Total GPCR Internalization Assay
Código de producto. 17333879
Encompass
Change view
Click to view available options
Cantidad:
200 dp (2 x 96-pocillos)
Tamaño de la unidad:
1 set
17333879 200 dp (2 x 96-pocillos) 1 set
1 options
Código de producto. 17333879
Marca: DiscoverX 930838E3CP0M
The PathHunter™ eXpress PTGIR Total GPCR Internalization Assay measures PTGIR (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ PTGIR Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. prostaglandin I2) based internalization of PTGIR via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress PK-tagged PTGIR and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the PTGIR receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Entrada del ensayo GPCR
Para utilizar con (aplicación) Assay-Ready eXpress Kits
Cantidad 200 dp (2 x 96-pocillos)
Entrada diana PTGIR
Objetivo PTGIR