Learn More
DiscoverX PathHunter™ eXpress MC3R Total GPCR Internalization Assay
Descripción
The PathHunter™ MC3R Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. melanocyte stimulating hormone) based internalization of MC3R via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress PK-tagged MC3R and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the MC3R receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | GPCR |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | MC3R |
| Objetivo | MC3R |