Learn More
DiscoverX PathHunter™ eXpress KDR Activity Assay
Descripción
The PathHunter™ KDR Activity eXpress kit provides an easy-to-use cell based assay to study small molecule drug candidates targeting RTKs. This kit assesses RTK activation via detection of resulting KDR phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress a PK-tagged engineered fusion of the catalytic domain of KDR to the extracellular ligand binding domain of EGFR alongside an EA-tagged SH2 domain protein. Upon ligand induced receptor activation, cross-phosphorylation of the receptor leads to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | SH2 recruitment |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Agonista primario | Recombinant Human EGF |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | KDR |
| Línea de células | U2OS |
| Objetivo | KDR |