Learn More
DiscoverX PathHunter™ eXpress GPR85 U2OS β-Arrestin Orphan GPCR Assay
Código de producto. 17343139
Encompass
Change view
Click to view available options
Cantidad:
200 dp (2 x 96-pocillos)
Tamaño de la unidad:
1 set
17343139 200 dp (2 x 96-pocillos) 1 set
1 options
Código de producto. 17343139
Marca: DiscoverX 930409E3CP1M
The PathHunter™ eXpress GPR85 U2OS β-Arrestin Orphan GPCR Assay measures GPR85 (GPCR) activity via recruitment of β-Arrestin2. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ GPR85 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR85 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged GPR85 and EA-tagged β-Arrestin 2. Activation of GPR85 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Orphan receptor with unknown ligand.
Especificaciones
Entrada del ensayo GPCR
Para utilizar con (aplicación) Assay-Ready eXpress Kits
Formato 200 dp (2 x 96-well)
Cantidad 200 dp (2 x 96-pocillos)
Entrada diana GPR85
Línea de células U2OS
Objetivo GPR85