Learn More
DiscoverX PathHunter™ eXpress F2R (PAR1) CHO-K1 β-Arrestin GPCR Assay
Código de producto. 17362889
Encompass
Change view
Click to view available options
Cantidad:
1000 dp (10 x 96-pocillos)
Tamaño de la unidad:
1 set
17362889 1000 dp (10 x 96-pocillos) 1 set
1 options
Código de producto. 17362889 Proveedor DiscoverX N.º de proveedor 930278E2CP0L
The PathHunter™ eXpress F2R (PAR1) CHO-K1 β-Arrestin GPCR Assay measures F2R (PAR1) (GPCR) activity via recruitment of β-Arrestin2. eXpress kits contain all assay materials: cells, reagents, and plates.
The PathHunter™ F2R (PAR1) β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. proteases, TFLLRN) based activation of F2R GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged F2R and EA-tagged β-Arrestin 2. Activation of F2R stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Especificaciones
Entrada del ensayo GPCR
Para utilizar con (aplicación) Assay-Ready eXpress Kits
Formato 1,000 dp (10 x 96-well)
Cantidad 1000 dp (10 x 96-pocillos)
Entrada diana F2R (PAR1)
Línea de células CHO-K1
Objetivo F2R (PAR1)
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.