Learn More
DiscoverX PathHunter™ eXpress cINSRb Functional Assay
Descripción
The PathHunter™ eXpress cINSRb Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated cINSRb dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged cINSRb and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Cyno version of the protein.
Especificaciones
Especificaciones
| Entrada del ensayo | Receptor Phosphorylation |
| Para utilizar con (aplicación) | Assay-Ready eXpress Kits |
| Agonista primario | Insulin, Human |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | cINSRb |
| Línea de células | U2OS |
| Objetivo | cINSRb |