Learn More
DiscoverX PathHunter™ CHO-K1 mCCR4 β-Arrestin Cell Line
Descripción
The PathHunter™ mCCR4 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL22/MDC) based activation of mCCR4 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mCCR4 and EA-tagged β-Arrestin 2. Activation of mCCR4 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein.
Especificaciones
Especificaciones
| Entrada del ensayo | β-Arrestin recruitment |
| Línea de células | CHO-K1 |
| Para utilizar con (aplicación) | Cell Lines |
| Agonista primario | Recombinant Murine MDC (CCL22) |
| Cantidad | 2 viales |
| Entrada diana | mCCR4 |
| Número de acceso | NM_009916.2 |