Learn More
DiscoverX PathHunter™ CHO-K1 CNR1 Bioassay Kit
Descripción
The PathHunter™ CNR1 Bioassay kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This bioassay assesses ligand (e.g. CP 55,940) based activation of CNR1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged CNR1 and EA-tagged β-Arrestin 2. Activation of CNR1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Especificaciones
Especificaciones
| Entrada del ensayo | β-Arrestin recruitment |
| Para utilizar con (aplicación) | Assay-Ready Bioassay Kits |
| Cantidad | 200 dp (2 x 96-pocillos) |
| Entrada diana | CNR1 |
| Línea de células | CHO-K1 |
| Objetivo | CNR1 |