missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PASD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5113119
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PASD1 Polyclonal Antibody for Western Blot, ELISA
This gene encodes a protein that is thought to function as a transcription factor. The protein is a cancer-associated antigen that can stimulate autologous T-cell responses, and it is therefore considered to be a potential immunotherapeutic target for the treatment of various hematopoietic malignancies, including diffuse large B-cell lymphoma.
Especificaciones
| PASD1 | |
| Polyclonal | |
| Unconjugated | |
| PASD1 | |
| Cancer/testis antigen 63; circadian clock protein PASD1; CT63; Gm1141; OXTES1; OX-TES-1; PAS domain containing 1; PAS domain-containing protein 1; PASD1; predicted gene 1141 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 139135 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IV76 | |
| PASD1 | |
| Recombinant Human Circadian clock protein PASD1 protein (301-600AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido