missing translation for 'onlineSavingsMsg'
Learn More

Parthenolide, MedChemExpress

Código de producto. 16464767
Encompass_Preferred
Change view
Click to view available options
Cantidad:
50 mg
100 mg
200 mg
Unit Size:
100 miligramos
200 miligramos
50 miligramos
Product Code. Cantidad unitSize
16464767 50 mg 50 miligramos
16409006 100 mg 100 miligramos
16401496 200 mg 200 miligramos
3 options
This item is not returnable. View return policy

Product Code. 16464767

Brand: Medchem Express HYN0141

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Parthenolide is a sesquiterpene lactone found in the medicinal herb Feverfew. Parthenolide exhibits anti-inflammatory activity by inhibiting NF-κB activation; also inhibits HDAC1 protein without affecting other class I/II HDACs.

Parthenolide is a sesquiterpene lactone found in the medicinal herb Feverfew. Parthenolide exhibits anti-inflammatory activity by inhibiting NF-κB activation

Specifications

Nombre del producto químico o material Parthenolide
CAS 20554-84-1
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C15H20O3
Cantidad 50 mg
Sinónimo (-)-Parthenolide
Información de solubilidad DMSO : ≥ 100 mg/mL (402.71 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C/C1=C\CC[C@@](C)(O2)[C@H]2[C@@H](OC(C3=C)=O)[C@H]3CC1
Peso molecular (g/mol) 248.32
Formula Weight (peso de la fórmula) 248.32
Porcentaje de pureza 97.11%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.