missing translation for 'onlineSavingsMsg'
Learn More

PARP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16842155 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Unit Size:
150 microlitros
20 microlitros
Product Code. Cantidad unitSize
16842155 20 μL 20 microlitros
16832155 150 μL 150 microlitros
2 options
This item is not returnable. View return policy

Product Code. 16842155

Brand: Proteintech 229991AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody was generated against the N-terminal region of human PARP1 and it recognizes the full-length as well as the cleavage of the PARP1.

Poly ADP-Ribose Polymerase (PARP) uses nicotinamide adenine dinucleotide (oxidized form) NAD as a substrate to catalyse the transfer of ADP-ribose to a variety of nuclear protein acceptors. Proteolysis of PARP to its stable 85kDa fragment is an early marker of programmed cell death (apoptosis) and is mediated by the caspase CPP32 protein. Cleavage occurs between Adp216 and Gly217, a site in PARP conserved across species.

Specifications

Antígeno PARP1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Clasificación Polyclonal
Concentración 0.31 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica PARP1
N.º de referencia del gen P09874, P11103
Alias de gen ADPRT, ADPRT 1, ADPRT1, pADPRT 1, PARP, PARP 1, PARP1, poly (ADP ribose) polymerase 1, Poly [ADP ribose] polymerase 1, Poly[ADP ribose] synthase 1, PPOL
Símbolos de los genes Parp1
Especie del huésped Rabbit
Inmunógeno PARP1 Fusion Protein Ag19173
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11545, 142
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.