missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAR2 Recombinant Rabbit Monoclonal Antibody (23GB5850), Invitrogen™

Descripción
Coagulation factor II (thrombin) receptor-like 1 (F2RL1) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL1 is also a member of the protease-activated receptor family. It is activated by trypsin, but not by thrombin. It is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. The F2RL1 gene contains two exons and is widely expressed in human tissues. The predicted protein sequence is 83% identical to the mouse receptor sequence.
Especificaciones
Especificaciones
| Antígeno | PAR2 |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 23GB5850 |
| Concentración | 0.381 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | F2rl1 |
| N.º de referencia del gen | P55085, P55086, Q63645 |
| Alias de gen | coagulation factor II (thrombin) receptor-like 1; coagulation factor II receptor-like 1; F2R like trypsin receptor 1; F2RL1; Gpcr11; Gpr11; G-protein coupled receptor 11; PAR2; PAR-2; protease-activated receptor 2; Protease-activated receptor-2; proteinase-activated receptor 2; Proteinase-activated receptor 2, alternate cleaved 1; Proteinase-activated receptor 2, alternate cleaved 2; proteinase-activated receptor-2; Proteinase-activated receptor-2 G protein-coupled receptor 11; Proteinase-activated receptor-2, G protein-coupled receptor 11; thrombin receptor-like 1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?