missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAPOLG Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA565147
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: KQLHHYLPAEI LQKKKKQSLS DVNRSSGGLQ SKRLSLDSSC LDSSRDTDNG TPFNSPASKS DSPSVGETER N Highest antigen sequence identity to the following orthologs - mouse 78%, rat 77%.
This gene overlaps with the gene for cysteineendopeptidase AUT-like 4 in a head-to-tail orientation.
Especificaciones
| PAPOLG | |
| Polyclonal | |
| Unconjugated | |
| PAPOLG | |
| 9630006B20Rik; FLJ11805; FLJ12972; FLJ13482; FLJ14187; hypothetical protein MGC63762; MGC133307; MGC133308; neo-PAP; Neo-poly(A) polymerase; nuclear poly(A) polymerase gamma; PAP2; PAPG; PAP-gamma; Papolg; Poly(A) polymerase gamma; Poly(A) polymerase gamma-like protein; polynucleotide adenylyltransferase gamma; Signal recognition particle RNA-adenylating enzyme; SRP 3'-adenylating protein; SRP RNA 3' adenylating enzyme/pap2; SRP RNA 3'-adenylating enzyme; SRP RNA-adenylating enzyme; zgc:63762 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BWT3 | |
| PAPOLG | |
| Recombinant Human PAPOLG. Recombinant protein control fragment (Product #RP-104660). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu