Learn More
STEMCELL Technologies Papaína
Código de producto. p-7217397
Encompass_Preferred
Click to view available options
Cantidad:
25 mg
100 mg
Tamaño de la unidad:
100 miligramos
25 miligramos
Código de producto. 17119281
Marca: STEMCELL Technologies 07465
For digestion of the extracellular matrix of cartilage
  • Papain is a cysteine protease consisting of a single polypeptide containing three disulfide bridges
  • Native crystalline papain is unreactive until acted upon by mild reducing agents, e.g. , cysteine, sulfide, or sulfite, and therefore likely exists as a zymogen
  • Papain has a wide specificity with a preference towards arginine, lysine, and phenylalanine
  • This enzyme degrades protein substrates such as the intercellular matrices of cartilage more extensively than pancreatic proteases and is typically less damaging and more effective than other proteases for tissue dissociation applications (Huettner & Baugham; Lam) and has also been used for the digestion of neural tissue (Fasano et al.)
Especificaciones
Descripción Dopaminergic Neurons; Mesenchymal Cells, PSC-Derived; Neurons; Pancreatic Cells
Cantidad 25 mg
Sinónimo Papainase; Papaya peptidase I
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.