Learn More
PANX1 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16890384 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Unit Size:
150 microlitros
20 microlitros
Product Code. Cantidad unitSize
16890384 20 μL 20 microlitros
16880384 150 μL 150 microlitros
2 options
This item is not returnable. View return policy
Product Code. 16890384
Brand: Proteintech 125951AP20UL
This item is not returnable. View return policy
Rabbit Polyclonal Antibody
The pannexin gene family encodes a second class of putative gap junction proteins and are highly conserved in invertebrates and mammals. Pannexins (Panx) are four-pass transmembrane proteins that oligomerize to form large pore ion and metabolite-permeable channels. Pannexin-1 (PANX1) and Pannexin-3 are closely related, while Pannexin-2 is a more distant relation. PANX1 is a transmembrane protein that forms a mechanosensitive ATP-permeable channel between adjacent cells and in the endoplasmic reticulum. PANX1 may play a role as a Ca2+ -leak channel to regulate ER Ca2+ homeostasis and regulates neural stem and progenitor cell proliferation.
Specifications
Antígeno PANX1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Clasificación Polyclonal
Concentración 0.14 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica PANX1
N.º de referencia del gen Q96RD7
Alias de gen MRS1, pannexin 1, PANX1, PX1, UNQ2529
Símbolos de los genes PANX1
Especie del huésped Rabbit
Inmunógeno PANX1 Fusion Protein Ag3174
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 24145
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG