Learn More
Palm Barcode Scanner CR3600 Dark Grey with Bluetooth and Charging Station
Descripción
Brady labels and label printers, benchtop or handheld, offer a full range of capabilities and options to provide high quality, cost-effective printing on a complete line of materials.
Brady offers bar code scanners that can meet the needs of a variety of tracking applications, including basic data entry, electronics, lab sample ID, shipping and receiving, UID Initiatives, WIP and many more.
Brady offers bar code scanners that can meet the needs of a variety of tracking applications, including basic data entry, electronics, lab sample ID, shipping and receiving, UID Initiatives, WIP and many more.
Especificaciones
Especificaciones
| Anchura (métrico) | 130 mm |
| Altura (métrico) | 36.00 mm |
| Fondo (métrico) | 51.00 mm |
| Dimensiones (L x An) | 36 x 130 mm |
| Descripción | CR3600 dark grey Palm scanner with bluetooth, battery and a charging station with embedded modem and a 1M USB charge cable |
| Resolución | 960 x 640 |
| Peso (métrico) | 0.15 kg |
| Certificaciones/conformidad | WEEE; CE; FCC; RoHs |